Product Details

SNP ID
rs201132768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:20369580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGTAAGGAGGCGGCGGCGGCGG[A/C]GGAGGAGGAAGCAGCCCCGGGAGCA
Phenotype
MIM: 123805
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PDE3A PubMed Links

Gene Details

Gene
PDE3A
Gene Name
phosphodiesterase 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000921.4 336 Missense Mutation GAG,GCG E99A NP_000912.3
NM_001244683.1 336 Intron NP_001231612.1
XM_017019420.1 336 Missense Mutation GAG,GCG E99A XP_016874909.1
XM_017019421.1 336 Intron XP_016874910.1

View Full Product Details