Product Details

SNP ID
rs200954465
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:2820744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGGTGGACAGCCTCTCAGTGAC[G/T]CTGGGGCCACTGGGTCTTCCTGAAC
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ITFG2 PubMed Links

Gene Details

Gene
ITFG2
Gene Name
integrin alpha FG-GAP repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018463.3 707 Silent Mutation ACG,ACT T189T NP_060933.3
XM_005253707.4 707 Silent Mutation ACG,ACT T189T XP_005253764.1
XM_005253709.4 707 Silent Mutation ACG,ACT T12T XP_005253766.1
XM_017019641.1 707 Intron XP_016875130.1
Gene
NRIP2
Gene Name
nuclear receptor interacting protein 2
There are no transcripts associated with this gene.

View Full Product Details