Product Details

SNP ID
rs200884864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:9833434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTAATAAGGACCTTTGTGCTTCG[A/G]GATCTGCAGACCAGACAGGTATGTC
Phenotype
MIM: 605029
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLRF1 PubMed Links

Gene Details

Gene
KLRF1
Gene Name
killer cell lectin like receptor F1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291822.1 571 Intron NP_001278751.1
NM_001291823.1 571 Intron NP_001278752.1
NM_016523.2 571 Missense Mutation AGA,GGA R106G NP_057607.1
XM_017019414.1 571 Missense Mutation AGA,GGA R106G XP_016874903.1
XM_017019415.1 571 UTR 3 XP_016874904.1

View Full Product Details