Product Details

SNP ID
rs200864066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:118068490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTTTGGCAACTCCTCTCTCTCA[C/T]GTGCTCCCTCCTGTTCCCCTACTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VSIG10 PubMed Links

Gene Details

Gene
VSIG10
Gene Name
V-set and immunoglobulin domain containing 10
There are no transcripts associated with this gene.

Gene
WSB2
Gene Name
WD repeat and SOCS box containing 2
There are no transcripts associated with this gene.

View Full Product Details