Product Details
- SNP ID
-
rs200888450
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:122511179 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCGCTGTTCTTGTAATTTCTTTGC[C/T]CTTTCCAACCTGCAAAATTAATTTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RSRC2
PubMed Links
Gene Details
- Gene
- RSRC2
- Gene Name
- arginine and serine rich coiled-coil 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_023012.5 |
895 |
Silent Mutation |
AGA,AGG |
R245R |
NP_075388.2 |
XM_005253601.1 |
895 |
Silent Mutation |
AGA,AGG |
R245R |
XP_005253658.1 |
XM_005253602.2 |
895 |
Silent Mutation |
AGA,AGG |
R197R |
XP_005253659.1 |
XM_005253604.1 |
895 |
Silent Mutation |
AGA,AGG |
R186R |
XP_005253661.1 |
XM_011538688.1 |
895 |
Silent Mutation |
AGA,AGG |
R245R |
XP_011536990.1 |
XM_011538689.1 |
895 |
Silent Mutation |
AGA,AGG |
R186R |
XP_011536991.1 |
XM_017019830.1 |
895 |
Silent Mutation |
AGA,AGG |
R245R |
XP_016875319.1 |
XM_017019831.1 |
895 |
Silent Mutation |
AGA,AGG |
R197R |
XP_016875320.1 |
XM_017019832.1 |
895 |
Silent Mutation |
AGA,AGG |
R13R |
XP_016875321.1 |
XM_017019833.1 |
895 |
Silent Mutation |
AGA,AGG |
R13R |
XP_016875322.1 |
View Full Product Details