Product Details
- SNP ID
-
rs201008493
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:10825662 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACATTCGATTGCATCTGCCTGTTG[C/T]GTCTCCAAAGGGAAATGATTAAAAA
- Phenotype
-
MIM: 604791
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TAS2R10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12307411] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TAS2R10
- Gene Name
- taste 2 receptor member 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_023921.1 |
608 |
Missense Mutation |
CAC,CGC |
H203R |
NP_076410.1 |
View Full Product Details