Product Details

SNP ID
rs201056424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6329795 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTCACTGTCTAGGCTCTGTGGC[C/T]TGTGGGCGCTGTCCTCCCACTTCTG
Phenotype
MIM: 611743 MIM: 191190
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLEKHG6 PubMed Links

Gene Details

Gene
PLEKHG6
Gene Name
pleckstrin homology and RhoGEF domain containing G6
There are no transcripts associated with this gene.

Gene
TNFRSF1A
Gene Name
TNF receptor superfamily member 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001065.3 1343 Missense Mutation AAG,AGG K347R NP_001056.1

View Full Product Details