Product Details

SNP ID
rs200045476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53269974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTATGAGAGCTGCCAGTTCTTCCT[C/T]TCAGGCCTGGAACGAGGCACCAAGA
Phenotype
MIM: 604143
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESPL1 PubMed Links

Gene Details

Gene
ESPL1
Gene Name
extra spindle pole bodies like 1, separase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012291.4 1123 Silent Mutation CTC,CTT L344L NP_036423.4
XM_006719705.3 1123 Silent Mutation CTC,CTT L344L XP_006719768.1
XM_011539024.2 1123 Silent Mutation CTC,CTT L344L XP_011537326.1
XM_011539025.2 1123 Silent Mutation CTC,CTT L344L XP_011537327.1
XM_017020253.1 1123 Silent Mutation CTC,CTT L19L XP_016875742.1

View Full Product Details