Product Details

SNP ID
rs200092271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:130163289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCATGTGCGAGCAGGCCCGGCTCA[A/G]GTGCTCCCCGATTATGGAGCAGTTC
Phenotype
MIM: 606147
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FZD10 PubMed Links

Gene Details

Gene
FZD10
Gene Name
frizzled class receptor 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007197.3 831 Missense Mutation AAG,AGG K116R NP_009128.1
Gene
FZD10-AS1
Gene Name
FZD10 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details