Product Details

SNP ID
rs200191564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:129074042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGCGGTGAAGGTGGTAAATTTTA[C/T]CCTTTTCCTTTTTGAGGTAGGGGAT
Phenotype
MIM: 611257
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM132D PubMed Links

Gene Details

Gene
TMEM132D
Gene Name
transmembrane protein 132D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133448.2 3461 Missense Mutation ATA,GTA I1045V NP_597705.2

View Full Product Details