Product Details

SNP ID
rs200319393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56642744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTACATCTTGACCTTCTTGGTCTC[C/T]GAAGTATTCAGCCACAGTCAGCCCA
Phenotype
MIM: 102910 MIM: 605682
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP5B PubMed Links

Gene Details

Gene
ATP5B
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001686.3 985 Missense Mutation AGA,GGA R294G NP_001677.2
Gene
BAZ2A
Gene Name
bromodomain adjacent to zinc finger domain 2A
There are no transcripts associated with this gene.

Gene
SNORD59A
Gene Name
small nucleolar RNA, C/D box 59A
There are no transcripts associated with this gene.

Gene
SNORD59B
Gene Name
small nucleolar RNA, C/D box 59B
There are no transcripts associated with this gene.

View Full Product Details