Product Details

SNP ID
rs200156270
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7323285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGAAATTAAACCAGGTTCAATG[A/G]GGAAAGGAATGCTGCCCTATGATGT
Phenotype
MIM: 614360 MIM: 606079
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACSM4 PubMed Links

Gene Details

Gene
ACSM4
Gene Name
acyl-CoA synthetase medium-chain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080454.1 1177 Missense Mutation AGG,GGG R393G NP_001073923.1
Gene
CD163L1
Gene Name
CD163 molecule like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297650.1 1177 Intron NP_001284579.1
NM_174941.5 1177 Intron NP_777601.2
XM_011520616.1 1177 Intron XP_011518918.1
XM_011520617.2 1177 Intron XP_011518919.1
XM_011520618.2 1177 Intron XP_011518920.1
XM_011520619.1 1177 Intron XP_011518921.1
XM_011520620.1 1177 Intron XP_011518922.1
XM_011520621.2 1177 Intron XP_011518923.1

View Full Product Details