Product Details
- SNP ID
-
rs200488834
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:76358707 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAGTCTGCAATAAATATTTTTACC[C/T]TCACTTCCAGAGGAGTCTTGAATGT
- Phenotype
-
MIM: 606736
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OSBPL8
PubMed Links
Gene Details
- Gene
- OSBPL8
- Gene Name
- oxysterol binding protein like 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001003712.1 |
3011 |
Silent Mutation |
GAA,GAG |
E769E |
NP_001003712.1 |
NM_001319652.1 |
3011 |
Silent Mutation |
GAA,GAG |
E769E |
NP_001306581.1 |
NM_001319653.1 |
3011 |
Silent Mutation |
GAA,GAG |
E808E |
NP_001306582.1 |
NM_001319655.1 |
3011 |
Silent Mutation |
GAA,GAG |
E786E |
NP_001306584.1 |
NM_020841.4 |
3011 |
Silent Mutation |
GAA,GAG |
E811E |
NP_065892.1 |
XM_005268621.4 |
3011 |
Silent Mutation |
GAA,GAG |
E811E |
XP_005268678.1 |
XM_006719224.2 |
3011 |
Silent Mutation |
GAA,GAG |
E769E |
XP_006719287.1 |
XM_011537855.2 |
3011 |
Silent Mutation |
GAA,GAG |
E821E |
XP_011536157.1 |
XM_011537856.2 |
3011 |
Silent Mutation |
GAA,GAG |
E824E |
XP_011536158.1 |
XM_011537857.2 |
3011 |
Silent Mutation |
GAA,GAG |
E824E |
XP_011536159.1 |
XM_017018768.1 |
3011 |
Silent Mutation |
GAA,GAG |
E834E |
XP_016874257.1 |
XM_017018769.1 |
3011 |
Silent Mutation |
GAA,GAG |
E808E |
XP_016874258.1 |
View Full Product Details