Product Details

SNP ID
rs200517253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120581205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGAGGCTGCTCAGAACTCGGTCAT[C/T]GAGGCTTAGGCGGCAGCGGGGGTCG
Phenotype
MIM: 609992 MIM: 615998
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POP5 PubMed Links

Gene Details

Gene
POP5
Gene Name
POP5 homolog, ribonuclease P/MRP subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015918.3 105 Missense Mutation NP_057002.2
NM_198202.1 105 Missense Mutation NP_937845.1
XM_011538441.1 105 Missense Mutation XP_011536743.1
Gene
RNF10
Gene Name
ring finger protein 10
There are no transcripts associated with this gene.

View Full Product Details