Product Details

SNP ID
rs200668464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122702264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGGCCATTAACGCCTCTGGAGC[A/G]CCTCTGGTTTTGTTGGGGTCCCCTG
Phenotype
MIM: 609163
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HCAR2 PubMed Links

Gene Details

Gene
HCAR2
Gene Name
hydroxycarboxylic acid receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177551.3 1094 Silent Mutation GGC,GGT G340G NP_808219.1

View Full Product Details