Product Details

SNP ID
rs200742427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53713884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGACTCGTCCTCTGAGTCTGT[C/T]AGAGCTGCCGGGCAGCCTGTAGGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CALCOCO1 PubMed Links

Gene Details

Gene
CALCOCO1
Gene Name
calcium binding and coiled-coil domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143682.1 1837 Silent Mutation CTA,CTG L451L NP_001137154.1
NM_020898.2 1837 Silent Mutation CTA,CTG L536L NP_065949.1
XM_011538601.1 1837 Silent Mutation CTA,CTG L536L XP_011536903.1
XM_011538602.1 1837 Silent Mutation CTA,CTG L536L XP_011536904.1
XM_011538603.1 1837 Silent Mutation CTA,CTG L536L XP_011536905.1
XM_011538604.1 1837 Silent Mutation CTA,CTG L536L XP_011536906.1
XM_017019707.1 1837 Silent Mutation CTA,CTG L536L XP_016875196.1
XM_017019708.1 1837 Silent Mutation CTA,CTG L213L XP_016875197.1
XM_017019709.1 1837 Silent Mutation CTA,CTG L213L XP_016875198.1

View Full Product Details