Product Details

SNP ID
rs200774200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:11133737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCATATTTGAATGGTACATTGCAC[G/T]CCTCAATTTGATCTTCCAAGTCACG
Phenotype
MIM: 168730 MIM: 613963
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRH1 PubMed Links

Gene Details

Gene
PRH1
Gene Name
proline rich protein HaeIII subfamily 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291314.1 508 Intron NP_001278243.1
NM_001291315.1 508 Intron NP_001278244.1
Gene
PRH1-PRR4
Gene Name
PRH1-PRR4 readthrough
There are no transcripts associated with this gene.

Gene
PRH1-TAS2R14
Gene Name
PRH1-TAS2R14 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316893.1 508 Intron NP_001303822.1
Gene
TAS2R30
Gene Name
taste 2 receptor member 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001097643.1 508 Missense Mutation AGT,CGT S170R NP_001091112.1

View Full Product Details