Product Details

SNP ID
rs199781088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49587606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCAGGCTTTTGTGGCAGGAGGAC[G/T]ACACGTCCAGTGTCAACAGCCGGGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM186B PubMed Links

Gene Details

Gene
FAM186B
Gene Name
family with sequence similarity 186 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006719626.2 Intron XP_006719689.1
XM_006719627.3 Intron XP_006719690.1
XM_011538797.2 Intron XP_011537099.1
Gene
LOC100422296
Gene Name
Parkinsonism associated deglycase pseudogene
There are no transcripts associated with this gene.

View Full Product Details