Product Details
- SNP ID
-
rs200000346
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:71941710 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTGAAGAATGAAGTTGGTGGATTG[A/G]TAAAAGCACTGAGGCTCTTTCAGGT
- Phenotype
-
MIM: 607478
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TPH2
PubMed Links
Gene Details
- Gene
- TPH2
- Gene Name
- tryptophan hydroxylase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173353.3 |
373 |
Missense Mutation |
ATA,GTA |
I78V |
NP_775489.2 |
View Full Product Details