Product Details
- SNP ID
-
rs199668545
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:53504737 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAGGCTGGCGGTTGCCGGAGTAC[A/C]CAGTGACCCAGGAGTCTGGGCCAGC
- Phenotype
-
MIM: 606371
MIM: 600447
MIM: 604643
MIM: 605053
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ATF7
PubMed Links
Gene Details
- Gene
- ATF7
- Gene Name
- activating transcription factor 7
There are no transcripts associated with this gene.
- Gene
- LOC100652999
- Gene Name
- uncharacterized LOC100652999
There are no transcripts associated with this gene.
- Gene
- MAP3K12
- Gene Name
- mitogen-activated protein kinase kinase kinase 12
There are no transcripts associated with this gene.
- Gene
- NPFF
- Gene Name
- neuropeptide FF-amide peptide precursor
There are no transcripts associated with this gene.
- Gene
- TARBP2
- Gene Name
- TARBP2, RISC loading complex RNA binding subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004178.4 |
642 |
Missense Mutation |
ACA,CCA |
T158P |
NP_004169.3 |
NM_134323.1 |
642 |
Missense Mutation |
ACA,CCA |
T179P |
NP_599150.1 |
NM_134324.2 |
642 |
Missense Mutation |
ACA,CCA |
T158P |
NP_599151.2 |
XM_005269114.1 |
642 |
Missense Mutation |
ACA,CCA |
T158P |
XP_005269171.1 |
XM_005269115.2 |
642 |
Missense Mutation |
ACA,CCA |
T158P |
XP_005269172.1 |
XM_005269117.1 |
642 |
Missense Mutation |
ACA,CCA |
T179P |
XP_005269174.1 |
XM_005269120.4 |
642 |
Missense Mutation |
ACA,CCA |
T35P |
XP_005269177.1 |
XM_005269122.3 |
642 |
Missense Mutation |
ACA,CCA |
T35P |
XP_005269179.1 |
XM_006719581.1 |
642 |
Missense Mutation |
ACA,CCA |
T80P |
XP_006719644.1 |
XM_011538712.2 |
642 |
Missense Mutation |
ACA,CCA |
T35P |
XP_011537014.1 |
XM_017019910.1 |
642 |
Missense Mutation |
ACA,CCA |
T158P |
XP_016875399.1 |
XM_017019911.1 |
642 |
Missense Mutation |
ACA,CCA |
T35P |
XP_016875400.1 |
View Full Product Details