Product Details
- SNP ID
-
rs199864668
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:44520114 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTCTTGGTGATGTCATTGCGGATG[A/G]TGTCAGCCTGGCAAGGGTCTGTGAC
- Phenotype
-
MIM: 602320
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NELL2
PubMed Links
Gene Details
- Gene
- NELL2
- Gene Name
- neural EGFL like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001145107.1 |
2566 |
Missense Mutation |
ACC,ATC |
T814I |
NP_001138579.1 |
NM_001145108.1 |
2566 |
Missense Mutation |
ACC,ATC |
T764I |
NP_001138580.1 |
NM_001145109.1 |
2566 |
Missense Mutation |
ACC,ATC |
T763I |
NP_001138581.1 |
NM_001145110.1 |
2566 |
Missense Mutation |
ACC,ATC |
T787I |
NP_001138582.1 |
NM_006159.2 |
2566 |
Missense Mutation |
ACC,ATC |
T764I |
NP_006150.1 |
XM_005268905.3 |
2566 |
Missense Mutation |
ACC,ATC |
T764I |
XP_005268962.1 |
XM_011538396.1 |
2566 |
Missense Mutation |
ACC,ATC |
T764I |
XP_011536698.1 |
XM_017019341.1 |
2566 |
Missense Mutation |
ACC,ATC |
T819I |
XP_016874830.1 |
XM_017019342.1 |
2566 |
Missense Mutation |
ACC,ATC |
T769I |
XP_016874831.1 |
XM_017019343.1 |
2566 |
Missense Mutation |
ACC,ATC |
T769I |
XP_016874832.1 |
XM_017019344.1 |
2566 |
Missense Mutation |
ACC,ATC |
T769I |
XP_016874833.1 |
View Full Product Details