Product Details

SNP ID
rs199818434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48568538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCAGTAGTCAATTCCTTTAATA[C/T]CCAGGATCTTCTTGGCACACATTAT
Phenotype
MIM: 149750
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LALBA PubMed Links

Gene Details

Gene
LALBA
Gene Name
lactalbumin alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002289.2 451 Missense Mutation GAT,GGT D116G NP_002280.1
XM_006719395.3 451 Missense Mutation GAT,GGT D123G XP_006719458.1

View Full Product Details