Product Details

SNP ID
rs201579033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57607090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCGGCCCCTCCAAATGGGCTCT[A/G]CCTAGCCCGGAAGGCTCTCAAGGGG
Phenotype
MIM: 610215 MIM: 613142
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF25 PubMed Links

Gene Details

Gene
ARHGEF25
Gene Name
Rho guanine nucleotide exchange factor 25
There are no transcripts associated with this gene.

Gene
DTX3
Gene Name
deltex E3 ubiquitin ligase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286245.1 919 Missense Mutation TAC,TGC Y79C NP_001273174.1
NM_001286246.1 919 Missense Mutation TAC,TGC Y76C NP_001273175.1
NM_178502.3 919 Missense Mutation TAC,TGC Y76C NP_848597.1
XM_005268697.1 919 Missense Mutation TAC,TGC Y79C XP_005268754.1
XM_005268698.1 919 Missense Mutation TAC,TGC Y79C XP_005268755.1
XM_005268700.1 919 Missense Mutation TAC,TGC Y79C XP_005268757.1
XM_005268703.1 919 Missense Mutation TAC,TGC Y76C XP_005268760.1
XM_011538022.2 919 Missense Mutation TAC,TGC Y76C XP_011536324.1
Gene
PIP4K2C
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 gamma
There are no transcripts associated with this gene.

View Full Product Details