Product Details

SNP ID
rs201721140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6829866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTTCGCCTCCTCAGTTGAAGA[A/G]GCTGGATCTGGCAGCTGCGGCAGCA
Phenotype
MIM: 186940 MIM: 610342
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD4 PubMed Links

Gene Details

Gene
CD4
Gene Name
CD4 molecule
There are no transcripts associated with this gene.

Gene
GPR162
Gene Name
G protein-coupled receptor 162
There are no transcripts associated with this gene.

Gene
P3H3
Gene Name
prolyl 3-hydroxylase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014262.4 574 Missense Mutation AAG,AGG K169R NP_055077.2

View Full Product Details