Product Details

SNP ID
rs201701567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:25204207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCAGAGAAGATCAAAGAACTTAT[G/T]GCACAGGGCGAATTTGTAATGAAAG
Phenotype
MIM: 616906 MIM: 190070
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CASC1 PubMed Links

Gene Details

Gene
CASC1
Gene Name
cancer susceptibility candidate 1
There are no transcripts associated with this gene.

Gene
KRAS
Gene Name
KRAS proto-oncogene, GTPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004985.4 1748 Intron NP_004976.2
NM_033360.3 1748 Intron NP_203524.1
XM_006719069.3 1748 Intron XP_006719132.1
XM_011520653.2 1748 Intron XP_011518955.1
XM_017019292.1 1748 Intron XP_016874781.1
XM_017019293.1 1748 Intron XP_016874782.1
Gene
LYRM5
Gene Name
LYR motif containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001660.2 1748 Intron NP_001001660.2
XM_005253319.4 1748 Intron XP_005253376.1
XM_005253320.4 1748 Intron XP_005253377.1
XM_017018850.1 1748 Missense Mutation ATG,ATT M56I XP_016874339.1

View Full Product Details