Product Details

SNP ID
rs201820175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:33376847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGGCATTATGGTGTGGAAGGTG[G/T]TTTAGGAGAAGGGCAGGATCCTTGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SYT10 PubMed Links

Gene Details

Gene
SYT10
Gene Name
synaptotagmin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198992.3 1213 Missense Mutation ACA,CCA T519P NP_945343.1
XM_011520644.2 1213 Missense Mutation ACA,CCA T338P XP_011518946.1

View Full Product Details