Product Details

SNP ID
rs201872894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57225070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGCTGGCCCGGGCAGGGGCAGCC[A/G]GGGCGTTGCCCGCGCAGCGCACCAA
Phenotype
MIM: 604637 MIM: 138450
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDUFA4L2 PubMed Links

Gene Details

Gene
NDUFA4L2
Gene Name
NDUFA4, mitochondrial complex associated like 2
There are no transcripts associated with this gene.

Gene
NXPH4
Gene Name
neurexophilin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007224.3 425 Missense Mutation AGG,GGG R84G NP_009155.1
XM_017018747.1 425 Missense Mutation AGG,GGG R84G XP_016874236.1
Gene
SHMT2
Gene Name
serine hydroxymethyltransferase 2
There are no transcripts associated with this gene.

View Full Product Details