Product Details

SNP ID
rs201782908
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:20369898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTGGCCGCCGCGACATGGCTGG[G/T]GCTGAGGCTGAGGCTGGGCGTCCTC
Phenotype
MIM: 123805
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PDE3A PubMed Links

Gene Details

Gene
PDE3A
Gene Name
phosphodiesterase 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000921.4 654 Missense Mutation GGG,GTG G205V NP_000912.3
NM_001244683.1 654 Intron NP_001231612.1
XM_017019420.1 654 Missense Mutation GGG,GTG G205V XP_016874909.1
XM_017019421.1 654 Intron XP_016874910.1

View Full Product Details