Product Details

SNP ID
rs201806935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:85805049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCTTTCATGCTTTTCTCCAAAT[C/T]ACACTTAACACTCTCTAAATTAGAG
Phenotype
MIM: 610383
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RASSF9 PubMed Links

Gene Details

Gene
RASSF9
Gene Name
Ras association domain family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005447.3 1300 Missense Mutation AAT,GAT N321D NP_005438.2
XM_011538988.2 1300 Missense Mutation AAT,GAT N300D XP_011537290.1

View Full Product Details