Product Details

SNP ID
rs202013500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:27780818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCACTTCCACGAGGTGCTGTGCA[A/T]GCCCCAGTTCCACCTCCTGGGGTCT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KLHL42 PubMed Links

Gene Details

Gene
KLHL42
Gene Name
kelch like family member 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020782.1 862 Missense Mutation AAG,ATG K163M NP_065833.1
XM_017019698.1 862 Missense Mutation AAG,ATG K163M XP_016875187.1
Gene
MANSC4
Gene Name
MANSC domain containing 4
There are no transcripts associated with this gene.

View Full Product Details