Product Details

SNP ID
rs202133075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:107688698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGTGCACGCACCCAGGCACGCC[C/G]AAGAGAGCCCCTGGAGGATGGTGAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PWP1 PubMed Links

Gene Details

Gene
PWP1
Gene Name
PWP1 homolog, endonuclein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317962.1 239 Missense Mutation CCA,CGA P10R NP_001304891.1
NM_001317963.1 239 UTR 5 NP_001304892.1
NM_007062.2 239 Missense Mutation CCA,CGA P72R NP_008993.1

View Full Product Details