Product Details

SNP ID
rs202133918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49349111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGACACCCCCTCTGAGTCCCATTC[A/G]CTTTGCTGCCCAGGTGATAGTTGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC22 PubMed Links

Gene Details

Gene
DNAJC22
Gene Name
DnaJ heat shock protein family (Hsp40) member C22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304944.1 981 Missense Mutation CAC,CGC H80R NP_001291873.1
NM_024902.3 981 Missense Mutation CAC,CGC H80R NP_079178.2
XM_005269155.2 981 Missense Mutation CAC,CGC H80R XP_005269212.1
XM_005269156.2 981 Missense Mutation CAC,CGC H80R XP_005269213.1
XM_005269157.2 981 Missense Mutation CAC,CGC H80R XP_005269214.1

View Full Product Details