Product Details
- SNP ID
-
rs202111268
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52823120 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTAGTCACGCAGCAGCCGTGTCAG[A/G]TCCTCCTTGGCCTGGTGCAGGGCCA
- Phenotype
-
MIM: 123940
MIM: 611160
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KRT4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73102423] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KRT4
- Gene Name
- keratin 4
There are no transcripts associated with this gene.
- Gene
- KRT79
- Gene Name
- keratin 79
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175834.2 |
1296 |
Silent Mutation |
GAC,GAT |
D421D |
NP_787028.1 |
View Full Product Details