Product Details
- SNP ID
-
rs202019064
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:6570887 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCCTACCTGCTGTGGGGTAGGTTC[A/G]GGTGCCCGGTTTGCCAGGCGGCTGA
- Phenotype
-
MIM: 603277
MIM: 164031
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CHD4
PubMed Links
Gene Details
- Gene
- CHD4
- Gene Name
- chromodomain helicase DNA binding protein 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001273.3 |
5807 |
Silent Mutation |
CCC,CCT |
P1901P |
NP_001264.2 |
NM_001297553.1 |
5807 |
Silent Mutation |
CCC,CCT |
P1894P |
NP_001284482.1 |
XM_005253668.3 |
5807 |
Silent Mutation |
CCC,CCT |
P1894P |
XP_005253725.1 |
XM_006718958.1 |
5807 |
Silent Mutation |
CCC,CCT |
P1929P |
XP_006719021.1 |
XM_006718959.1 |
5807 |
Silent Mutation |
CCC,CCT |
P1904P |
XP_006719022.1 |
XM_006718960.1 |
5807 |
Silent Mutation |
CCC,CCT |
P1903P |
XP_006719023.1 |
XM_006718961.2 |
5807 |
Silent Mutation |
CCC,CCT |
P1897P |
XP_006719024.1 |
XM_006718962.1 |
5807 |
Silent Mutation |
CCC,CCT |
P1891P |
XP_006719025.1 |
XM_017018725.1 |
5807 |
Silent Mutation |
CCC,CCT |
P1903P |
XP_016874214.1 |
XM_017018726.1 |
5807 |
Silent Mutation |
CCC,CCT |
P1902P |
XP_016874215.1 |
XM_017018727.1 |
5807 |
Silent Mutation |
CCC,CCT |
P1901P |
XP_016874216.1 |
XM_017018728.1 |
5807 |
Silent Mutation |
CCC,CCT |
P1900P |
XP_016874217.1 |
XM_017018729.1 |
5807 |
Silent Mutation |
CCC,CCT |
P1895P |
XP_016874218.1 |
XM_017018730.1 |
5807 |
Silent Mutation |
CCC,CCT |
P1889P |
XP_016874219.1 |
XM_017018731.1 |
5807 |
Silent Mutation |
CCC,CCT |
P1888P |
XP_016874220.1 |
XM_017018732.1 |
5807 |
Silent Mutation |
CCC,CCT |
P1884P |
XP_016874221.1 |
XM_017018733.1 |
5807 |
Silent Mutation |
CCC,CCT |
P1882P |
XP_016874222.1 |
XM_017018734.1 |
5807 |
Silent Mutation |
CCC,CCT |
P1881P |
XP_016874223.1 |
- Gene
- NOP2
- Gene Name
- NOP2 nucleolar protein
There are no transcripts associated with this gene.
View Full Product Details