Product Details

SNP ID
rs202187350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56206648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGACCCTGGGGCCAGCTACGCTC[A/G]TGGGCATTTTCAATCAGCTCGTTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF41 PubMed Links

Gene Details

Gene
RNF41
Gene Name
ring finger protein 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242826.1 1184 Silent Mutation CAC,CAT H251H NP_001229755.1
NM_005785.3 1184 Silent Mutation CAC,CAT H251H NP_005776.1
NM_194358.2 1184 Silent Mutation CAC,CAT H180H NP_919339.1
NM_194359.2 1184 Silent Mutation CAC,CAT H251H NP_919340.1
XM_005268561.4 1184 Silent Mutation CAC,CAT H180H XP_005268618.1
XM_011537734.2 1184 Silent Mutation CAC,CAT H180H XP_011536036.1
XM_011537735.2 1184 Silent Mutation CAC,CAT H180H XP_011536037.1

View Full Product Details