Product Details

SNP ID
rs202050803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52171461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACATTTCGGTGATTTTGATCAC[A/G]GGGCCTTTGCTGCCCTTCTTCTTTC
Phenotype
MIM: 611161
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT80 PubMed Links

Gene Details

Gene
KRT80
Gene Name
keratin 80
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081492.1 1429 UTR 3 NP_001074961.1
NM_182507.2 1429 Silent Mutation CCC,CCT P432P NP_872313.2
XM_005268676.3 1429 Silent Mutation CCC,CCT P467P XP_005268733.1

View Full Product Details