Product Details

SNP ID
rs201366396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55551601 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTATGATCGTTATGTGGCCATCTG[C/T]AAACCCTTGCATTACCTGACTATTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR6C4 PubMed Links

Gene Details

Gene
OR6C4
Gene Name
olfactory receptor family 6 subfamily C member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005494.1 375 Silent Mutation TGC,TGT C125C NP_001005494.1

View Full Product Details