Product Details

SNP ID
rs201160699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14774505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGGGAAACAGGGCGGCAAAGTGC[A/G]AGCAAAGGCCAAATCCCGCTCCTCC
Phenotype
MIM: 615069
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C12orf60 PubMed Links

Gene Details

Gene
C12orf60
Gene Name
chromosome 12 open reading frame 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175874.3 170 Intron NP_787070.2
XM_005253322.4 170 Intron XP_005253379.1
XM_011520568.2 170 Intron XP_011518870.1
XM_011520569.2 170 Intron XP_011518871.1
XM_017018871.1 170 Intron XP_016874360.1
XM_017018872.1 170 Intron XP_016874361.1
XM_017018873.1 170 Intron XP_016874362.1
XM_017018874.1 170 Intron XP_016874363.1
Gene
H2AFJ
Gene Name
H2A histone family member J
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177925.3 170 Missense Mutation CAA,CGA Q12R NP_808760.1
Gene
HIST4H4
Gene Name
histone cluster 4, H4
There are no transcripts associated with this gene.

Gene
LOC105369669
Gene Name
uncharacterized LOC105369669
There are no transcripts associated with this gene.

View Full Product Details