Product Details

SNP ID
rs201162712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49870607 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAATATACTCCTCAGGGCTCAGC[A/G]AGTGGCGTCGGTTACCCATCAGCAA
Phenotype
MIM: 604306
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAIM2 PubMed Links

Gene Details

Gene
FAIM2
Gene Name
Fas apoptotic inhibitory molecule 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012306.3 1140 Missense Mutation TCG,TTG S283L NP_036438.2
XM_005268730.3 1140 Missense Mutation TCG,TTG S241L XP_005268787.1
XM_017019040.1 1140 Missense Mutation TCG,TTG S283L XP_016874529.1

View Full Product Details