Product Details

SNP ID
rs201282200
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:66129031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGAATACTTTTAAGCCTGCTGG[A/C]CTCCTTTGGGGCATTCTTTTTTCTC
Phenotype
MIM: 616998 MIM: 616874
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
LLPH PubMed Links
Additional Information
For this assay, SNP(s) [rs1179916] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LLPH
Gene Name
LLP homolog, long-term synaptic facilitation
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032338.3 Intron NP_115714.1
Gene
LLPH-AS1
Gene Name
LLPH antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
TMBIM4
Gene Name
transmembrane BAX inhibitor motif containing 4
There are no transcripts associated with this gene.

View Full Product Details