Product Details

SNP ID
rs201305943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121843771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGTCTTCAGCTTCTCCCGCAGT[G/T]GTTTGTAGTACGTGGAGGGAACAGA
Phenotype
MIM: 609695
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HPD PubMed Links

Gene Details

Gene
HPD
Gene Name
4-hydroxyphenylpyruvate dioxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171993.1 933 Missense Mutation CAA,CCA Q259P NP_001165464.1
NM_002150.2 933 Missense Mutation CAA,CCA Q298P NP_002141.1

View Full Product Details