Product Details

SNP ID
rs201193016
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:107692825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTTTCTTACAGATGCTGAGACT[C/T]TTGGTGAATCTCTCTTGGGTCTTAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PWP1 PubMed Links

Gene Details

Gene
PWP1
Gene Name
PWP1 homolog, endonuclein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317962.1 355 Missense Mutation CTT,TTT L49F NP_001304891.1
NM_001317963.1 355 UTR 5 NP_001304892.1
NM_007062.2 355 Missense Mutation CTT,TTT L111F NP_008993.1

View Full Product Details