Product Details

SNP ID
rs201286832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53482830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGATCCCCAGCTCCGCCTGTG[C/G]CCCCCCGGCCCCGGGACCCTAGTGC
Phenotype
MIM: 600447 MIM: 601210
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAP3K12 PubMed Links

Gene Details

Gene
MAP3K12
Gene Name
mitogen-activated protein kinase kinase kinase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193511.1 2172 Missense Mutation GCC,GGC A658G NP_001180440.1
NM_006301.3 2172 Missense Mutation GCC,GGC A625G NP_006292.3
XM_005269138.3 2172 Missense Mutation GCC,GGC A658G XP_005269195.1
XM_005269140.2 2172 Intron XP_005269197.1
XM_006719588.3 2172 Missense Mutation GCC,GGC A658G XP_006719651.1
XM_011538725.2 2172 Missense Mutation GCC,GGC A658G XP_011537027.1
XM_017019956.1 2172 Missense Mutation GCC,GGC A625G XP_016875445.1
Gene
PCBP2
Gene Name
poly(rC) binding protein 2
There are no transcripts associated with this gene.

View Full Product Details