Product Details

SNP ID
rs201172877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54500550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTCAGCAACATTTAGGACCAGT[A/C]CATCGTGAAAAAGCCGAGATAATTA
Phenotype
MIM: 141180
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NCKAP1L PubMed Links

Gene Details

Gene
NCKAP1L
Gene Name
NCK associated protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184976.1 181 Silent Mutation GTA,GTC V27V NP_001171905.1
NM_005337.4 181 Silent Mutation GTA,GTC V77V NP_005328.2

View Full Product Details