Product Details

SNP ID
rs201287830
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:69251279 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGTGGGTAAAGGAGCAGCACCA[A/C]ATGTTGTCTATACATATACTGGAAA
Phenotype
MIM: 604979
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CPSF6 PubMed Links

Gene Details

Gene
CPSF6
Gene Name
cleavage and polyadenylation specific factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300947.1 321 Missense Mutation AAT,CAT N71H NP_001287876.1
NM_007007.2 321 Missense Mutation AAT,CAT N71H NP_008938.2
XM_005268588.2 321 Missense Mutation AAT,CAT N71H XP_005268645.1
XM_005268590.2 321 Missense Mutation AAT,CAT N71H XP_005268647.1

View Full Product Details