Product Details

SNP ID
rs202131204
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:51934766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGCCTGAAGTCATCAGATGTAC[G/T]GCTCCTCATCCCTGCCATTCAGGAG
Phenotype
MIM: 606882
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATP7B PubMed Links

Gene Details

Gene
ATP7B
Gene Name
ATPase copper transporting beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000053.3 3924 Missense Mutation CAG,CCG Q1463P NP_000044.2
NM_001005918.2 3924 Missense Mutation CAG,CCG Q1256P NP_001005918.1
NM_001243182.1 3924 Missense Mutation CAG,CCG Q1352P NP_001230111.1
XM_005266423.2 3924 Missense Mutation CAG,CCG Q1431P XP_005266480.1
XM_005266424.4 3924 Missense Mutation CAG,CCG Q1431P XP_005266481.1
XM_005266427.2 3924 Missense Mutation CAG,CCG Q1385P XP_005266484.1
XM_005266428.1 3924 Missense Mutation CAG,CCG Q1379P XP_005266485.1
XM_005266430.4 3924 Missense Mutation CAG,CCG Q1463P XP_005266487.1
XM_005266431.3 3924 Missense Mutation CAG,CCG Q1451P XP_005266488.1
XM_005266432.2 3924 Missense Mutation CAG,CCG Q1301P XP_005266489.1
XM_006719837.3 3924 Missense Mutation CAG,CCG Q1431P XP_006719900.1
XM_006719838.1 3924 Missense Mutation CAG,CCG Q735P XP_006719901.1
XM_006719839.1 3924 Missense Mutation CAG,CCG Q674P XP_006719902.1
XM_011535117.2 3924 Missense Mutation CAG,CCG Q1431P XP_011533419.1
XM_011535118.1 3924 Missense Mutation CAG,CCG Q1418P XP_011533420.1
XM_011535119.1 3924 Missense Mutation CAG,CCG Q1402P XP_011533421.1
XM_011535122.2 3924 Missense Mutation CAG,CCG Q1019P XP_011533424.1
XM_017020627.1 3924 Missense Mutation CAG,CCG Q1431P XP_016876116.1
XM_017020628.1 3924 Missense Mutation CAG,CCG Q1019P XP_016876117.1

View Full Product Details