Product Details

SNP ID
rs200797698
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:41192914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCATTCCACTTCCTTCAACTTAAC[G/T]CCAGTAATAGGGTCTCGTCCCCCCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KBTBD7 PubMed Links

Gene Details

Gene
KBTBD7
Gene Name
kelch repeat and BTB domain containing 7
There are no transcripts associated with this gene.

Gene
LOC101929140
Gene Name
uncharacterized LOC101929140
There are no transcripts associated with this gene.

View Full Product Details