Product Details

SNP ID
rs201257355
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:110615716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCGGAACTGCCGTCGCCGGCGC[C/G]GTCGTTGTCGCATTGCTCTCGGCCG
Phenotype
MIM: 615910
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NAXD PubMed Links

Gene Details

Gene
NAXD
Gene Name
NAD(P)HX dehydratase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242881.1 133 Silent Mutation GCC,GCG A14A NP_001229810.1
NM_001242882.1 133 Intron NP_001229811.1
NM_001242883.1 133 Intron NP_001229812.1
NM_018210.3 133 Silent Mutation GCC,GCG A14A NP_060680.2

View Full Product Details