Product Details

SNP ID
rs201258088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:25097320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTAGCTCAGCGCAAAGAAGAGCG[C/T]CAGGCTTACCTCACTAACGAGTATA
Phenotype
MIM: 604680
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PABPC3 PubMed Links

Gene Details

Gene
PABPC3
Gene Name
poly(A) binding protein cytoplasmic 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030979.2 Intron NP_112241.2

View Full Product Details