Product Details
- SNP ID
-
rs200806993
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:26046814 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTCTGTGACCTTGTTGGGGGCGCC[C/T]GGGCCCCTGAGTTGGCGCTGGAGCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SHISA2
PubMed Links
Gene Details
- Gene
- SHISA2
- Gene Name
- shisa family member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001007538.1 |
672 |
Missense Mutation |
CAG,CGG |
Q196R |
NP_001007539.1 |
View Full Product Details